Learn More
Granzyme K Mouse anti-Human, Unconjugated, Clone: 1C3A4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67272-1-IG-150UL
 View more versions of this product
Product Code. 16839805
  • 4836.10 NOK / 150µL
Description
The Granzyme K antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Granzyme K. This antibody recognizes human antigen. The Granzyme K antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
Specifications
Granzyme K
Monoclonal
500 μg/mL
Mouse
Primary
Human
Immunohistochemistry, Immunofluorescence, ELISA
1C3A4
Unconjugated
150 μL
3003
IgG2b