missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granzyme B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NB100-684SS
This item is not returnable.
View return policy
Description
Granzyme B Polyclonal antibody specifically detects Granzyme B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications
| Granzyme B | |
| Polyclonal | |
| Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen 1:10-1:500 | |
| C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTSGL1, granzyme B, GranzymeB, GZMB, HLP, SECT | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 3002 | |
| Human, Mouse, Rat | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| PBS (pH 7.4), 0.2% BSA, Tween-20 with 0.05% Sodium Azide | |
| GZMB | |
| Synthetic peptide: EIIGGHEAKPHSRPYMAYL, corresponding to amino acids 20-38 of Human Granzyme B (N-terminus). | |
| 0.1 mL | |
| Primary | |
| This reacts with a 32 kDa protein. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction