Learn More
3M™ Graffiti Remover 3000
Product Code. p-7221322
Change view
Click to view available options
Volume (Metric):
0.5 L
5 L
Unit Size:
1 bottle
Product Code. Volume (Metric) unitSize
17207640 0.5 L 1 bottle
17297630 5 L 1 bottle
2 options
Product Code. 17207640
Brand: 3M™ 7100030784
Graffiti Remover 3000
  • Graffiti remover for plastic, painted or varnished surfaces - graffiti remover is effective against aerosol spray paint, permanent markers, lipstick, crayon and emulsion paint
  • The solvent’s liquid form allows for quick penetration and rapid breakdown of graffiti
  • Does not contain NEP (N-Ethyl-pyrrolidone) or NMP (N-Methyl-2-pyrrolidone) used in other graffiti removers, which can pose a significant risk to health
  • Has little odour, enhancing user comfort and efficiency – unlike traditional chemical removers which emit powerful odours and harmful vapours
  • Low environmental impact as the product does not contain aromatic, aliphatic or chlorinated solvents
TRUSTED_SUSTAINABILITY
Specifications
Product Type Graffiti Remover
Volume (Metric) 0.5 L