Learn More
3M™ Graffiti Remover 3000
Product Code. 17207640
Change view
Click to view available options
Volume (Metric):
0.5 L
5 L
Unit Size:
1 bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
17207640 0.5 L 1 bottle
17297630 5 L 1 bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17207640 Supplier 3M™ Supplier No. 7100030784
  • ONLINE EXCLUSIVE PRICE
    1955.00 NOK
    1871.83 NOK / 1 bottles
    Save 83.17 NOK
    4% Off
View all promotions
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
Graffiti Remover 3000
  • Graffiti remover for plastic, painted or varnished surfaces - graffiti remover is effective against aerosol spray paint, permanent markers, lipstick, crayon and emulsion paint
  • The solvent’s liquid form allows for quick penetration and rapid breakdown of graffiti
  • Does not contain NEP (N-Ethyl-pyrrolidone) or NMP (N-Methyl-2-pyrrolidone) used in other graffiti removers, which can pose a significant risk to health
  • Has little odour, enhancing user comfort and efficiency – unlike traditional chemical removers which emit powerful odours and harmful vapours
  • Low environmental impact as the product does not contain aromatic, aliphatic or chlorinated solvents
TRUSTED_SUSTAINABILITY
Specifications
Product Type Graffiti Remover
Volume (Metric) 0.5 L