Learn More
3M™ Graffiti Remover 1500
Product Code. 17159340
Change view
Click to view available options
Volume (Metric):
500 mL
5 L
Unit Size:
4 bottles
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
17159340 500 mL 4 bottles
17169340 5 L 4 bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17159340 Supplier 3M™ Supplier No. 7000082043
  • ONLINE EXCLUSIVE PRICE
    3120.00 NOK
    2043.96 NOK / Pack of 4
    Save 1076.04 NOK
    34% Off
 Restricted Item
Estimated Shipment: 11-01-2027
A safe and simple way to remove unsightly graffiti from plastic, painted, or varnished surfaces.
  • Graffiti remover for plastic, painted or varnished surfaces
  • Effective against aerosol spray paint, permanent markers, lipstick, crayon, and emulsion paint
  • The solvent’s liquid form allows for quick penetration and rapid breakdown of graffiti
  • Does not contain NEP (N-Ethyl-pyrrolidone) or NMP (N-Methyl-2-pyrrolidone) used in other graffiti removers, which can pose a significant risk to health
  • Has minimal odor, enhancing user comfort and efficiency unlike traditional chemical removers which emit powerful odors and harmful vapors
  • Low environmental impact since the product does not contain aromatic, aliphatic, or chlorinated solvents
TRUSTED_SUSTAINABILITY
Specifications
Product Type Graffiti Remover
Form Liquid
For Use With (Application) Graffiti Removal
For Use With (Equipment) Plastic, Painted or Varnished Surfaces
Volume (Metric) 500 mL
compliance-icons
Product Identifier
  • 3M Graffiti Remover 1500
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-(2-Ethoxyethoxy)ethanol, benzyl alcohol, (2-Methoxymethylethoxy)propanol, Fatty acids, C16-18, 3-b