Description
- PP, highly transparent
- Category B (DIN 12681 / ISO 6706)
- Embossed blue graduation with rings at the main points
- Shatterproof
- Adjusted to “In”
Specifications
Specifications
| Capacity (Metric) | 2000 mL |
| Material | Polypropylene |
| Certifications/Compliance | Category B DIN 12681/ISO 6706 |
| Scale | Embossed |
| Diameter (Metric) | 97 mm |
| Includes | Embossed blue graduation with rings at the main points |
| Quantity | 1 |
| Graduations | 20 mL |
| Autoclavable | Autoclavable |
| Shape | Round |
| Show More |