Learn More
Hirschmann™ Graduated Pipettes with Piston
Description
- Graduated to tip
- Available in a range of capacities
Specifications
Specifications
| Graduated | Yes |
| Graduation Color | Amber |
| Graduation Type | Amber graduations permanently with silver diffusion color |
| Quantity | 1 Each |
| Volume (Metric) | 5 mL |
| Length (Metric) | 360 mm |
| Type | Graduated Pipet |