Learn More
Hirschmann™ Graduated Pipettes with Piston
Provides volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Graduated Pipettes with Piston come with amber stain graduation.
Brand:  Hirschmann™ 1190175
Product Code. 10024740
  • ONLINE EXCLUSIVE PRICE
    1020.00 NOK
    897.30 NOK / Pack of 1
    Save 122.70 NOK
    12% Off
Estimated Shipment: 14-12-2026
Description
  • Graduated to tip
  • Available in a range of capacities
Specifications
Specifications
Yes
0.2 mL
1 Each
530 mm
Amber
Amber graduations permanently with silver diffusion color
50 mL
Graduated Pipette