Learn More
Hecht Karl™ Graduated Glass Cylinder
Specifications
Specifications
| Description | Low Form |
| Height (Metric) | 90 mm |
| Type | Graduated Cylinder |
| Material | Glass |
| Capacity (Metric) | 10 mL |