Learn More
KARTELL Graduated Burette
Product Code. 10632322
Change view
Click to view available options
Capacity (Metric):
10 mL
50 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10632322 10 mL Each
10748294 50 mL Each
2 options
Product Code. 10632322
Brand: KARTELL 0199700
Only null left
Add to basket
Add to basket
This Graduated Burette is highly precise and ideal for routine work.
  • High precison plastic burettes (according to DIN 12.700 standards - Class B), particularly useful for routine work to eliminate the danger of glass breakages.
  • The combination of different plastic materials (PMMA, PTFE, FEP) provides good resistance to chemicals.
  • The stopcock assembly is moulded in one piece, with FEP key; requiring no lubricant and providing a leak-proof seal.
Specifications
Capacity (Metric) 10 mL
Length (Metric) 580 mm
Material Acrylic
Graduations 0.1 mL
Length (English) 22.83 in.
Stopcock Material PTFE
Type Graduated Buret