Description
Applications:
- Cleaning and drying of, glass containers, tubes, flasks, microscopic plates, bottles
Specifications
Specifications
| Length (Metric) | 220 mm |
| Width (Metric) | 400 mm |
| Dimensions (L x W) | 220 x 400 mm |
| Format | Sheet |
| For Use With (Application) | Cleaning and drying of, Glass containers, tubes, flasks, microscopic plates, bottles in laboratories and hospitals |
| Packaging Quantity | 500 Pk. |
| Grade | 605 |