Learn More
Sartorius™ Creped Surface Filter Discs Qualitative Filter Papers, Grade 5 H/N
Used for routine analyses like clarification and determination of substances
Brand: Sartorius™ FT-3-423-240
Additional Details : Weight : 0.10000kg
Description
- Made of refined pulp and cotton linters with an >95% α-cellulose content
- Ash content between 0.1–0.15% (grade 100/N < 0.1%)
- Wet-strengthened
Specifications
Disc | |
3μm/sec. | |
5 H/N | |
0.28 mm | |
240 mm |
Cotton Linters, Refined Pulp | |
Used for the rapid filtration of relatively coarse precipitates; because of their creped structure they provide a larger filtration area than smooth filter paper | |
100 Pack | |
Qualitative Filter Paper | |
240 mm |