missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Creped Surface Filter Discs Qualitative Filter Papers, Grade 5 H/N

Used for routine analyses like clarification and determination of substances

Brand:  Sartorius™ FT-3-423-150

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10206091

  • 225.00 NOK / Pack of 100
In Stock
to see stock.
Add to basket

Description

Description

  • Made of refined pulp and cotton linters with an >95% α-cellulose content
  • Ash content between 0.1–0.15% (grade 100/N < 0.1%)
  • Wet-strengthened
Specifications

Specifications

Disc
3μm/sec.
5 H/N
0.28 mm
150 mm
Cotton Linters, Refined Pulp
Used for the rapid filtration of relatively coarse precipitates; because of their creped structure they provide a larger filtration area than smooth filter paper
100 Pack
Qualitative Filter Paper
150 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers