Learn More
Sartorius Stedim Biotech™ Grade 39/N Creped Papers for Qualitative and Technical Analyses, Discs
Sartorius Stedim Biotech™ Grade 39/N, Creped Papers for Qualitative and Technical Analyses, Discs are suitable for analytical and technical purposes where a fast flow rate is desirable.
Brand:  Sartorius Stedim Biotech™ FT-3-483-380
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15982950
  • 2420.00 NOK / Pack of 100
Estimated Shipment: 01-07-2024
Description
  • Made of refined pulp and cotton linters with > 95% alpha-cellulose
  • Ash content between 0.1 and 0.15%
  • Wet-strengthened
  • Very fast filtering, wide-pore
Specifications
Specifications
180 g/m2
Cotton Linters, Refined Pulp
5 sec.
100/Pk.
380 mm
Disc
15 μm
39/N
0.65 mm