Learn More
Thermo Scientific™ Grade 25 Sapphire Spheres, Dia.: 1mm
Product Code. 11346019
Change view
Click to view available options
Quantity:
100 Pieces
25 Pieces
5 Pieces
Unit Size:
Pack of 100
Pack of 25
Pack of 5
Product Code. Quantity unitSize
11346019 100 Pieces Pack of 100
15431767 5 Pieces Pack of 5
11336019 25 Pieces Pack of 25
3 options
Product Code. 11346019
Brand: Thermo Scientific™ 042067.KN
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Sapphire Sphere
Diameter (Metric) 1 mm
Grade Grade 25
Quantity 100 Pieces