missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GPT Polyclonal antibody specifically detects GPT in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | GPT |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | AAT1alanine aminotransferase 1, ALT1, ALT1EC 2.6.1.2, Glutamate pyruvate transaminase 1, glutamic-alanine transaminase 1, Glutamic--alanine transaminase 1, glutamic-pyruvate transaminase (alanine aminotransferase), Glutamic--pyruvic transaminase 1, GPT1GPT 1 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human GPT (NP_005300.1).,, Sequence:, MASSTGDRSQAVRHGLRAKVLTLDGMNPRVRRVEYAVRGPIVQRALELEQELRQGVKKPFTEVIRANIGDAQAMGQRPITFLRQVLALCVNPDLLSSPNFPDDAKKRAERILQACGGHSLGAYSVSSGIQLIREDVARYIERRDGGIPADPNNVFLSTGASDAIVTVLKLLVAGEGHTRTGVLIPIPQYP |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?