Learn More
Invitrogen™ GPR68 Recombinant Rabbit Monoclonal Antibody (16H23L16)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ 702595
Description
This antibody is predicted to react with Monkey, Rat, Mouse, Horse. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
GPR68 (also known as OGR1 - ovarian cancer G-protein coupled receptor 1) is a proton-sensing receptor involved in pH homeostasis. It represents an osteoblastic pH sensor regulating the cell-mediated response to acidosis in bone. GPR68 mediates its action by association with G proteins that stimulate inositol phosphate (IP) production or CA2+ mobilization. The receptor is almost silent at pH 7.8 but fully activated at pH 6.8. Also found to serve as a tumor suppressor in prostate cancer. Mutations to the gene result in amelogenesis imperfecta, hypomaturation type 2A6.
Specifications
| GPR68 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q15743 | |
| GPR68 | |
| Peptide corresponding to human GPR68 [aa342-aa365]. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| 16H23L16 | |
| Unconjugated | |
| GPR68 | |
| BB131428; Brgr1; Brgrb; G protein-coupled receptor 68; GPR12A; Gpr68; G-protein coupled receptor 68; Ogr1; OGR-1; ovarian cancer G protein-coupled receptor, 1; ovarian cancer G-protein coupled receptor 1; Sphingosylphosphorylcholine receptor | |
| Rabbit | |
| Protein A | |
| RUO | |
| 8111 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.