missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR58/TAAR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24609-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Not expressed in the pons, thalamus, hypothalamus, hippocampus, caudate, putamen, frontal cortex, basal forebrain, midbrain or liver.Specifications
GPR58/TAAR2 | |
Polyclonal | |
Unconjugated | |
TAAR2 | |
G protein coupled receptor 58, GPR58, TAAR2, TaR 2, Trace amine receptor 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
209512, 294121, 9287 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5QD17, Q5QD25, Q9P1P5 | |
TAAR2 | |
GPR58/TAAR2 Fusion Protein Ag18939 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |