missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR35 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55248-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to GPR35.
GPCR GPR116 is a member of the G protein coupled receptor (GPCR) family. Members of this protein family contain seven putative transmembrane domains and may mediate signaling processes to the interior of the cell via activation of heterotrimeric G proteins.Specifications
GPR35 | |
Polyclonal | |
Unconjugated | |
GPR35 | |
G protein coupled receptor 35, GPR35, KYNA receptor, Kynurenic acid receptor | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2859 | |
-20°C | |
Liquid |
Flow Cytometry, Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9HC97 | |
GPR35 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |