Learn More
Thermo Scientific™ GeneBLAzer™ GPR24-Gqi5-NFAT-bla CHO-K1 Cells
GeneBLAzer™ GPR24-Gqi5-NFAT-bla CHO-K1 cells contain the human Melanin-concetrating hormone receptor (GPR24) stably integrated into the GeneBLAzer™ Gqi5-NFAT-bla CHO-K1 cell line.
Brand:  Thermo Scientific™ K1730
Additional Details : Weight : 0.01000kg
Product Code. 10003833
  • 134500.00 NOK / Each
Estimated Shipment: 14-08-2024
Description
GeneBLAzer™ GPR24-Gqi5-NFAT-bla CHO-K1 cells contain the human Melanin-concetrating hormone receptor (GPR24) stably integrated into the GeneBLAzer™ Gqi5-NFAT-bla CHO-K1 cell line. GeneBLAzer™ Gqi5-NFAT-bla CHO-K1 cells (Cat. No. K1725) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element and the Chimeric G protein, Gqi5.
The GeneBLAzer™ GPR24-Gqi5-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of MCH. In addition, GeneBLAzer™ GPR24-Gqi5-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
Specifications
Specifications
BC021146.1
Dividing Cells
Fluorescence
GPR24
Single Cell Clone Origin
2 x 106 Cells
MCHR1, GPR24
CHO-K1
GPCR
Cell-based beta lactamase reporter gene
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.
Includes:
• GeneBLAzer™ GPR24-Gqi5-NFAT-bla CHO-K1 cells
• GeneBLAzer™ GPR24-Gqi5-NFAT-bla CHO-K1 Assay Protocol
• Certificate of Analysis
The GeneBLAzer™ GPR24-Gqi5-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
FRET
GPR24-Gqi5-NFAT-bla CHO-K1 cells, GPR24-Gqi5-NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis
GeneBLAzer™
BLA (Beta-Lactamase)
Dividing Cells
GeneBLAzer™