Learn More
GPR177 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17950-1-AP-150UL
 View more versions of this product
Product Code. 16847964
  • 4836.10 NOK / 150µL
Description
The function remains unknown.
Specifications
Specifications
GPR177
Polyclonal
Unconjugated
WLS
C1orf139, EVI, G protein coupled receptor 177, GPR177, Protein wntless homolog, Sprinter, WLS, Wntless
Rabbit
Antigen Affinity Chromatography
RUO
362065, 68151, 79971
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5T9L3, Q6DID7, Q6P689
Wls
GPR177 Fusion Protein Ag12428
150 μL
Primary
Human, Mouse, Rat