missing translation for 'onlineSavingsMsg'
Learn More

GPR177 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16857964 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16857964 20 μL 20µL
16847964 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16857964

Brand: Proteintech 179501AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The function remains unknown.

Specifications

Antigen GPR177
Applications Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene WLS
Gene Accession No. Q5T9L3, Q6DID7, Q6P689
Gene Alias C1orf139, EVI, G protein coupled receptor 177, GPR177, Protein wntless homolog, Sprinter, WLS, Wntless
Gene Symbols Wls
Host Species Rabbit
Immunogen GPR177 Fusion Protein Ag12428
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362065, 68151, 79971
Target Species Mouse, Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.