Learn More
GPR125 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11912-1-AP-20UL
Description
The GPR125 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GPR125. This antibody recognizes human, pig, mouse antigen. The GPR125 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| GPR125 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 166647 | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 333 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Pig, Mouse |