missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPM6A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15044-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Neuronal membrane glycoprotein M6-a is a protein that in humans is encoded by the GPM6A gene. Involved in neuronal differentiation, including differentiation and migration of neuronal stem cells. Plays a role in neuronal plasticity and is involved in neurite and filopodia outgrowth, filopodia motility and probably synapse formation. GPM6A-induced filopodia formation involves mitogen-activated protein kinase (MAPK) and Src signaling pathways. May be involved in neuronal NGF-dependent Ca2+ influx. May be involved in regulation of endocytosis and intracellular trafficking of G-protein-coupled receptors (GPCRs); enhances internalization and recycling of mu-type opioid receptor.Specifications
GPM6A | |
Polyclonal | |
Unconjugated | |
GPM6A | |
glycoprotein M6A, GPM6, GPM6A, M6A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
234267, 2823, 306439 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P35802, P51674, Q812E9 | |
Gpm6a | |
GPM6A Fusion Protein Ag6915 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |