Description
This gene encodes a putative RNA-binding protein containing G-patch and KOW (Kyprides, Ouzounis, Woese) domains. The encoded protein interacts directly with protein kinase A and protein kinase X and is also found associated with the spliceosome.
Specifications
Specifications
| Antigen | GPKOW |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | GPKOW |
| Gene Accession No. | Q56A08, Q92917 |
| Gene Alias | G patch domain and KOW motifs, GPATC5, GPATCH5, GPKOW, Protein MOS2 homolog, Protein T54, T54 |
| Gene Symbols | GPKOW |
| Show More |