missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPI Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15171-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene belongs to the GPI family whose members encode multifunctional phosphoglucose isomerase proteins involved in energy pathways. The protein encoded by this gene is a dimeric enzyme that catalyzes the reversible isomerization of glucose-6-phosphate and fructose-6-phosphate. The protein functions in different capacities inside and outside the cell. In the cytoplasm, the gene product is involved in glycolysis and gluconeogenesis, while outside the cell it functions as a neurotrophic factor for spinal and sensory neurons. Defects in this gene are the cause of nonspherocytic hemolytic anemia and a severe enzyme deficiency can be associated with hydrops fetalis, immediate neonatal death and neurological impairment.Specifications
GPI | |
Polyclonal | |
Unconjugated | |
GPI | |
AMF, Autocrine motility factor, g6pi, Glucose 6 phosphate isomerase, glucose phosphate isomerase, GNPI, GPI, Neuroleukin, NLK, PGI, PHI, Phosphoglucose isomerase, Phosphohexose isomerase, SA 36, Sperm antigen 36 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14751, 2821, 292804 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Western Blot, Immunocytochemistry | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P06744, P06745, Q6P6V0 | |
Gpi | |
GPI Fusion Protein Ag7423 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |