Learn More
GP1BA Mouse, anti-Human, Unconjugated, Clone: 3G1F5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67857-1-IG-150UL
Description
GP1BA, also named as CD42b , is a 145-kDa surface membrane protein of platelets, participates in the formation of platelet plugs by binding to the A1 domain of vWF, which is already bound to the subendothelium. It has a heterodimer form which composed of GP1BA and GP1BB.
Specifications
| GP1BA | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, ELISA | |
| 3G1F5 | |
| Unconjugated | |
| 150 μL | |
| 2811 | |
| Liquid |