Learn More
GP1BA Mouse, anti-Human, Unconjugated, Clone: 3G1F5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67857-1-IG-150UL
 View more versions of this product
Product Code. 17221526
  • 4836.10 NOK / 150µL
Description
GP1BA, also named as CD42b , is a 145-kDa surface membrane protein of platelets, participates in the formation of platelet plugs by binding to the A1 domain of vWF, which is already bound to the subendothelium. It has a heterodimer form which composed of GP1BA and GP1BB.
Specifications
Specifications
GP1BA
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
3G1F5
Unconjugated
150 μL
2811