Learn More
A&D Weighing GP Series Precision Balances Internal Calibration Class II Approved
GREENER_CHOICE
Product Code. 16681122
Change view
Click to view available options
Capacity:
101 kg
101 kg/61 kg
12 kg
21 kg
31 kg
31 kg/6.1 kg
61 kg
Readability:
0.1 g
1 g
1 g/0.1 g (smart range)
10 g/1 g (smart range)
Unit Size:
Each
Product Code. Capacity Readability unitSize
16681122 12 kg 0.1 g Each
16601132 31 kg 0.1 g Each
16621132 31 kg/6.1 kg 1 g/0.1 g (smart range) Each
16651122 101 kg 1 g Each
16651132 61 kg 1 g Each
16671122 101 kg/61 kg 10 g/1 g (smart range) Each
16691122 21 kg 0.1 g Each
7 options
Product Code. 16681122
Brand: A&D Weighing GP12KEC
With capacities from 12 to 102 kg, these high precision industrial balances have a large weighing pan, and also feature IP65 dust and waterproof protection. SmartRange feature for improved precision, regardless of tare container weight.
  • EC Type approved
  • Swing arm design or separate display and platform design
  • IP65 protected
  • Durable aluminum alloy & stainless steel construction
  • Easy calibration with internal calibration mass
  • SHS response speed of only 1.5 seconds
  • GLP, GMP and ISO compliant
  • RS-232C interface
  • Comparator output and analog output (optional)
  • Large Vacuum Fluorescent display
  • Data memory function
  • Counting function with ACAI (Automatic Counting Accuracy Improvement)
  • Smart range function (GP-32K/102K/32KS)
  • Comparator function
  • Underhook weighing for thermal and magnetic samples as well as for density calculation
  • Animal weighing function
Specifications
Capacity 12 kg
Display Type VFD
Readability 0.1 g
Stabilization Time 1.5 seconds
Type Industrial Precision Balance