Learn More
Gosselin™ Straight Container
Product Code. 15408844
Change view
Click to view available options
Capacity (Metric):
60 mL
125 mL
180 mL
Closure Color:
Blue
Green
Red
Unit Size:
Pack of 264
Pack of 380
Pack of 600
Pack of 700
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Color Unit Size
15408844 180 mL Green Pack of 264
15448814 60 mL Green Pack of 700
15448824 60 mL Red Pack of 600
15458824 125 mL Blue Pack of 380
15488824 125 mL Green Pack of 380
15498844 180 mL Red Pack of 264
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15408844 Supplier Gosselin™ Supplier No. TP53C004
  • ONLINE EXCLUSIVE PRICE
    1745.00 NOK
    970.87 NOK / Pack of 264
    Save 774.13 NOK
    44% Off
View all promotions
Estimated Shipment: 23-12-2026
Corning™ Gosselin™ Straight Container, Specimen Vials, PP come in different sizes and quantities, and are available sterile and non-sterile.
  • Clear and break-resistant polypropylene (PP)
  • Available in different sizes and quantities
  • Color-coded screw caps
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Sterility Sterile
Capacity (Metric) 180 mL
Closure Color Green
Quantity 264/Pk.