Learn More
Corning™ 1 mL Straw Pipet, Polypropylene, Sterile
Decrease cross contamination risks using disposable pipets
Brand: Corning™ EPI24B-01
Additional Details : Weight : 1.00000kg
Description
- Product traceability on each bag
- Sterile 10-3
- Packaged individually in tamper evident single zip bags
Specifications
Polypropylene | |
6000/Cs. | |
2.2 mL | |
Straw Pipet |
Sterile | |
25/Bag | |
24 cm | |
Single-use |