Learn More
Corning™ Sterile Polyethylene Blender Bags
Product Code. 11787919
Change view
Click to view available options
Capacity (Metric):
400 mL
Description:
With full filter
With lateral filter
Without filter
Quantity:
50/Pk.
Unit Size:
Pack of 1500
Pack of 2500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Description Quantity Unit Size
11787919 400 mL With lateral filter 50/Pk. Pack of 1500
11777919 400 mL With full filter 50/Pk. Pack of 1500
11755085 400 mL Without filter 50/Pk. Pack of 2500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11787919 Supplier Corning™ Supplier No. BBAG04
  • ONLINE EXCLUSIVE PRICE
    11485.00 NOK
    6692.77 NOK / Pack of 1500
    Save 4792.23 NOK
    41% Off
View all promotions
Estimated Shipment: 15-12-2026
Universal bags for homogeneous suspension
  • Polyethylene (PE) bags
  • 80 to 3500 mL
  • With or without filter
  • Traceability on each filter bag
  • Sterile
Order Info
Use bag clip, Cat. No. 07-201-979, to close bags between sample preparation steps.
TRUSTED_SUSTAINABILITY
Specifications
Description With lateral filter
Color Natural
Material Polyethylene
Sterility Sterile
No. per Case 1500/Cs.
No. per Pack 500/Pk.
Quantity 50/Pk.
Capacity (Metric) 400 mL
Length (Metric) 300 mm
Width (Metric) 190 mm