Learn More
Gosselin™ Gosselin™ Round Base Tube
Click to view available options
Capacity (Metric):
2 mL
Unit Size:
Pack of 6000
Description
- Centrifugation up to 3000 g with a suitable adaptor in a balanced rotor
- Compatible with automation systems
- For laboratory use
- Polystyrene (PS) or Polypropylene (PP)
Specifications
Specifications
| Capacity (Metric) | 2 mL |
| Material | Polystyrene |
| Quantity | 1500/Pk., 6000/Cs. |