Learn More
Gosselin™ Polystrene Petri Dish
Description
- Appropriate for aseptic production
- Sterilized by irradiation
- Flat bottom
Specifications
Specifications
| Certifications/Compliance | EC N°1935/2004, ISO 11137 |
| Color | Natural |
| Material | Polystyrene |
| Shape | Round |
| Sterility | Sterile |
| Type | Petri Dish |
| Well Count | 1 |
| For Use With (Application) | For use in clinical, environment, industries, microbiology |
| Quantity | 1620 Carton |
| Vent | 0 |
| Show More |