Learn More
Gosselin™ Conical Containers
Product Code. 12612225
Click to view available options
Capacity (Metric):
200 mL
400 mL
1000 mL
Color:
Natural
White
Diameter (Metric):
62 mm
85 mm
105 mm
Closure Included:
No
Yes
Yes (Unassembled)
Unit Size:
Pack of 120
Pack of 220
Pack of 250
Pack of 460
Pack of 660
Product Code. 12612225
Brand: Gosselin™ PC100009
Provides safe and secure storage in the laboratory. Gosselin™ Conical Containers are manufactured from either clear or white polypropylene.
  • Shock resistant
  • Molded graduations outside on clear containers; molded internal graduations on white containers
TRUSTED_SUSTAINABILITY
Specifications
Product Type Container
Closure Included Yes
Graduated Yes
Material Polypropylene
Shape Conical, Cylindrical
Sterility Non-sterile
Capacity (Metric) 1000 mL
Certifications/Compliance EC N°1935/2004
Closure Type Snap Cap
Color Natural
Diameter (Metric) 105 mm
Height (Metric) 130 mm
Includes LDPE snap cap
No. per Case 120/Cs.