Learn More
Gosselin™ Inoculating L-Shaped Spreaders
GREENER_CHOICE
Product Code. 12908140
Change view
Click to view available options
Quantity:
Pack of 1000
Quantity per Carton : 2000
Quantity per Carton : 6000
Subpacks of 5
Unit Size:
Pack of 1000
Pack of 400
Pack of 6000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12908140 Pack of 1000 Pack of 1000
11821741 Quantity per Carton : 2000 Pack of 400
11836191 Subpacks of 5 Pack of 400
12622265 Quantity per Carton : 6000 Pack of 6000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12908140 Supplier Gosselin™ Supplier No. ETAR01
Estimated Shipment: 20-01-2027
Add to basket
Add to basket
For spreading over the surface of solid media in microbiological applications. Gosselin™ Inoculating L-Shaped Spreaders are manufactured from polystyrene.
  • Curved end
  • Sterile and disposable
  • Packed in tamper-evident ZIP bags
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
Certifications/Compliance EC N°1935/2004, ISO 11137
Color White
Length (Metric) 148 mm
Quantity Pack of 1000
Shape L-Shape
Material Polystyrene
Product Type Inoculating Spreader
Sterility Sterile
For Use With (Application) For Dairy, Environmental, Industrial, Microbiological Uses