Learn More
Gosselin™ Square HDPE Bottles
Product Code. p-8002239
Click to view available options
Closure Color:
Red
White
Diameter (Metric) Inner Neck:
22 mm
28 mm
29 mm
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Pack of 77
Product Code. 11703094
Brand: Gosselin™ FC51002
Only null left
Add to basket
Add to basket
Pre-dosed with sodium thiosulfate which inhibits the action of chlorine, bromine and ozone dissolved in water. Gosselin™ Square HDPE Bottles are manufactured from translucent HDPE.
  • Break resistant
  • With molded graduations
  • With either a screw cap with shaped seal or hinged cap with inner lip and safety ring
  • Sterilized by irradiation with peel-away sterilization certificate on the outer carton
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 500 mL
Color Translucent
Diameter (Metric) Inner Neck 29 mm
Includes Red LDPE hinged cap
Material HDPE
Length (Metric) 70mm
Quantity 100/Carton
Sterility Sterile
Width (Metric) 70mm
Closure Color Red
Certifications/Compliance EC N°1935/2004, ISO 11137
Closure Included Yes
Disposable Single-use
Mouth Narrow
Shape Square
Height (Metric) 148 mm
Closure Material LDPE
Closure Type Hinged Cap