Learn More
Gosselin™ Pocket ZIP Bags
Product Code. 11743254
Change view
Click to view available options
Color:
Blue
Natural
Yellow
Length (Metric):
170 mm
220 mm
300 mm
Unit Size:
Pack of 1000
Product Code. Color Length (Metric) unitSize
11743254 Natural 220 mm Pack of 1000
11713254 Blue 170 mm Pack of 1000
11723254 Natural 170 mm Pack of 1000
11793254 Natural 300 mm Pack of 1000
11887313 Yellow 170 mm Pack of 1000
5 options
Product Code. 11743254
Brand: Gosselin™ SP508
Only null left
Add to basket
Add to basket
Provides an excellent transport and storage option for solid samples. Gosselin™ Pocket ZIP Bags are nonsterile and manufactured from PE plastic.
  • Quick closure system
  • Single or double pocket
  • Available in a range of colors
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
For Use With (Application) Used in collection and transportation of solid samples and tubes
Material Polyethylene
Thickness (Metric) 50 μm
Quantity 2000 Carton
Length (Metric) 220 mm
Width (Metric) 160 mm