Learn More
Gosselin™ Pocket ZIP Bags
Description
- Quick closure system
- Single or double pocket
- Available in a range of colors
Specifications
Specifications
| Color | Blue |
| For Use With (Application) | Used in collection and transportation of solid samples and tubes |
| Material | Polyethylene |
| Thickness (Metric) | 50 μm |
| Quantity | 4000 Carton |
| Length (Metric) | 170 mm |
| Width (Metric) | 125 mm |