Description
- Light guide with eco-friendly Puravis fiber optics.
- Precise positioning offers complete control of light placement.
- Gooseneck branches positioned in opposite directions create near shadow-free illumination.
- Re-adjust with ease.
- For use with Cold light source KL300LED. Diameter: End tip: 6 mm, Lightsource adapter: 9 mm, Active Fiber optic bundle: 4.5 mm, Outer Jacketing: 6.5 mm
Specifications
Specifications
| Type | Gooseneck Single Light Guide |
| Description | Single Branch Fiber Optic Lightguide |
| Diameter (Metric) | 11 mm |
| For Use With (Application) | Stereo Microscopy Illumination |
| For Use With (Equipment) | Cold Light Source KL300LED |
| Length (Metric) | 500 mm |
| Material | Puravis Glass Optical Fibers |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?