Learn More
SCHOTT AG Lighting and Imaging™ Gooseneck Light Guide
Shop All Schott Products
Click to view available options
Type:
Gooseneck Double Light Guide
Gooseneck Single Light Guide
Gooseneck Triple Light Guide
Unit Size:
Each
Description
- Light guide with eco-friendly Puravis™ fiber optics.
- Precise positioning offers complete control of light placement.
- Gooseneck branches are positioned in opposite directions to create near shadow-free illumination.
- Re-adjust with ease.
- For use with cold light source KL1600LED, KL2500LED or KL1500HAL.
Specifications
Specifications
| Type | Gooseneck Triple Light Guide |
| Diameter (English) | 0.4 in. |
| Diameter (Metric) | 11 mm |
| Diameter (Metric) Inner | 4.5 mm |
| Dimensions (L x W x H) | 4.5 mm Fiber Bundle Diameter |
| For Use With (Application) | Stereo Microscopy Illumination |
| For Use With (Equipment) | Cold Light Source KL1600LED, KL2500LED or KL1500HAL |
| Length (English) | 23.6 in. |
| Length (Metric) | 600 mm |
| Length (Metric) Arm Reach | 600 mm |
| Show More |