Learn More
Gomisin G, MedChemExpress
Product Code. 16489138
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16489138
Brand: Medchem Express HYN0858
MedChemExpress Gomisin G is an ethanolic extract of the stems of Kadsura interior; exhibits potent anti-HIV activity with EC50 and therapeutic index (TI) values of 0.006 microgram/mL and 300, respectively.
Gomisin G is an ethanolic extract of the stems of Kadsura interior
Specifications
Chemical Name or Material Gomisin G
CAS 62956-48-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H32O9
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (93.18 mM)
SMILES COC1=C(OCO2)C2=CC([C@H](OC(C3=CC=CC=C3)=O)[C@](O)(C)[C@@H](C)C4)=C1C(C4=CC(OC)=C5OC)=C5OC
Molecular Weight (g/mol) 536.57
Formula Weight 536.57
Percent Purity 99.9%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.