missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Golgin-97 Monoclonal Antibody (CDF4), eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ 14-9767-37

 View more versions of this product

Product Code. 17160772

  • 15920.00 NOK / 2mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Description: The monoclonal antibody CDF4 recognizes human Golgin-97. Golgin-97, a GRIP domain protein member that was first identified as a Golgi complex autoantigen associated with Sjogren's syndrome. Golgin-97 is a peripheral membrane protein found on the trans face of the Golgi apparatus. The protein G family member, Arl1, mediates the binding of Golgin-97 to the Golgi membrane. Like all GRIP proteins, Golgin-97 functions in maintaining the integrity of the trans Golgi network. Golgin-97 has also been shown to function as a tethering molecule in the retrograde transport of proteins from the endosomes to the trans Golgi network. During poxvirus infection, Golgin-97 incorporates into virions and plays a role in viral replication and morphogenesis. Applications Reported: This CDF4 antibody has been reported for use in western blotting, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, immunohistochemical staining, and immunocytochemistry. Applications Tested: This CDF4 antibody has been tested by immunocytochemistry of fixed and permeabilized HeLa cells and can be used at less than or equal to 1 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. The Golgins are a family of proteins, of which the protein encoded by this gene is a member, that are localized to the Golgi. This encoded protein is associated with Sjogren's syndrome.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Golgin-97
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q92805
GOLGA1
Affinity chromatography
RUO
2800
4°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
CDF4
Unconjugated
GOLGA1
0710001G09Rik; 2210418B03Rik; AW107649; gap junction protein, alpha 4, 37kD; Golga1; golgi autoantigen, golgin subfamily a, 1; Golgi97; golgin A1; golgin subfamily A member 1; golgin-97
Mouse
2 mg
Primary
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.