missing translation for 'onlineSavingsMsg'
Learn More

GOLGA3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 21193-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16890815

  • 1281.92 NOK / 20µL
Estimated Shipment: 22-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a member of kinesin-like protein family. Proteins of this family are microtubule-dependent molecular motors that transport organelles within cells and move chromosomes during cell division. This protein is important for anaphase chromosome segregation and may be required to coordinate the onset of sister centromere separation.
Specifications

Specifications

GOLGA3
Polyclonal
Unconjugated
GOLGA3
GCP170, GOLGA3, Golgin 160, Golgin subfamily A member 3, MEA 2
Rabbit
Antigen Affinity Chromatography
RUO
269682, 2802
-20°C
Liquid
Western Blot, Immunocytochemistry, Immunofluorescence
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P55937, Q08378
Golga3
GOLGA3 Fusion Protein Ag15531
20 μL
Primary
Mouse, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers