Learn More
Thermo Scientific™ GC Injection Port Base Seals for Thermo Scientific™ and Agilent Instruments
Product Code. 10781947 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric) Inner:
0.8 mm
1.2 mm
For Use With (Equipment):
Dual Column Installation
Single Column Installation
Single Column Installation (Cross Version)
Quantity:
10 Pack
2 Pack
Product Type:
Gold Inlet Base Seal
Siltek Inlet Base Seal
Unit Size:
Pack of 10
Pack of 2
Product Code. Diameter (Metric) Inner For Use With (Equipment) Quantity Product Type unitSize
10781947 0.8 mm Single Column Installation 2 Pack Siltek Inlet Base Seal Pack of 2
10281665 0.8 mm Single Column Installation 10 Pack Gold Inlet Base Seal Pack of 10
10548605 0.8 mm Single Column Installation 2 Pack Gold Inlet Base Seal Pack of 2
10558605 1.2 mm Dual Column Installation 10 Pack Gold Inlet Base Seal Pack of 10
10703996 1.2 mm Dual Column Installation 2 Pack Gold Inlet Base Seal Pack of 2
10219174 0.8 mm Single Column Installation (Cross Version) 2 Pack Gold Inlet Base Seal Pack of 2
10371335 0.8 mm Single Column Installation (Cross Version) 10 Pack Gold Inlet Base Seal Pack of 10
10534655 0.8 mm Single Column Installation 10 Pack Siltek Inlet Base Seal Pack of 10
10575425 0.8 mm Single Column Installation (Cross Version) 2 Pack Siltek Inlet Base Seal Pack of 2
10417714 0.8 mm Single Column Installation (Cross Version) 10 Pack Siltek Inlet Base Seal Pack of 10
10 options
Product Code. 10781947
Brand: Thermo Scientific™ 290GA092
Only null left
Add to basket
Add to basket
Ensure optimal GC analysis by using the exceptional sealing and inert properties of Thermo Scientific™ GC Injection Port Base Seals.
Ensure optimal gas chromatography (GC) analysis by using exceptional sealing and inert properties of Thermo Scientific™ GC Injection Port Base Seals. Precision machined from high grade stainless steel, choose from gold-plated or Siltek-treated base seals. The chemically inert coated surface makes them suitable for analysis of active compounds such as pesticides, polychlorinated biphenyls (PCBs) and phenols. Use these GC Injection Port Base Seals with split/splitless injection ports of thermo Scientific™ TRACE™ 1300 Series GC and Agilent GC 5890/6890/7890 systems.
  • Precision machined to provide exceptional sealing properties
  • Reduces activity (especially useful for analysis of pesticide, PCBs, phenols, etc.)
  • High grade stainless steel provides reproducible seal
  • Direct equivalent to Agilent original part
  • Compatible with all Agilent GC 5890/6890/7890 split/splitless injection ports
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2 Pack
Product Type Siltek Inlet Base Seal
Diameter (Metric) Inner 0.8 mm
For Use With (Equipment) Single Column Installation