Learn More
Brand™ Sample Cups for COULTER COUNTER™, PS, with Cover, PE
Product Code. 10738181 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
20 mL
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10738181 20 mL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10738181 Supplier Brand™ Supplier No. 722055
  • ONLINE EXCLUSIVE PRICE
    1500.00 NOK
    1393.83 NOK / Pack of 1000
    Save 106.17 NOK
    7% Off
Estimated Shipment: 23-10-2026
Virgin Polystyrene (PS) sample cups for Coulter Counter with lids that are designed to fit many common analyzers.
Polystyrene, transparent. Lid PE.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 20 mL
Product Type Coulter Counter
Material Polystyrene