Learn More
AIRE Goat anti-Human, Mouse, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of AIRE.
Brand: Abnova PAB7187.100ug
Description
This gene encodes a transcriptional regulator that forms nuclear bodies and interacts with the transcriptional coactivator CBP. At least three splice variant mRNAs products have been described including one which results in a premature stop codon and a transcript predicted to be a candidate for nuclear-mediated decay (NMD). Defects in this gene cause the rare autosomal-recessive systemic autoimmune disease termed autoimmune polyendocrinopathy-candidiasis-ectodermal dystrophy (APECED). [provided by RefSeq
Sequence: C-KAKPPKKPESSAEQSpecifications
| AIRE | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:64000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| AIRE | |
| AIRE1/APECED/APS1/APSI/PGA1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 326 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of AIRE. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_000374.1;NP_000649.1 | |
| AIRE | |
| A synthetic peptide corresponding to human AIRE. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only