Learn More
Honeywell™ GO55 Carabiner
Product Code. 12615003
Change view
Click to view available options
For use with fall protection equipment.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12615003 For use with fall protection equipment.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12615003 Supplier Honeywell™ Supplier No. 1009249
  • ONLINE EXCLUSIVE PRICE
    713.44 NOK
    438.73 NOK / Pack of 1
    Save 274.71 NOK
    38% Off
Estimated Shipment: 12-04-2027
Steel carabiner for fall protection equipment that features a wide-opening hook.
  • Steel construction
  • Wide-opening hook
  • For fall protection equipment
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EC Category PPE: 3, QA: ISO 9001/2000, EC Attestation: 0082/036/160/09/00/0256
For Use With (Equipment) Fall protection equipment
Includes Opening: 55 mm
Strength Breaking strength: >2500 daN
Weight (Metric) 0.450 g
Width (Metric) 115 mm
Length (Metric) 218 mm
Material Steel