missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GO Protein alpha Antibody [DyLight 488], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
GO Protein alpha Polyclonal antibody specifically detects GO Protein alpha in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | GO Protein alpha |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DKFZp686O0962, G-ALPHA-o, GNAO, guanine nucleotide binding protein (G protein), alpha activating activitypolypeptide O, guanine nucleotide binding protein, alpha activating polypeptide O, guanine nucleotide-binding protein G(o) subunit alpha |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 60-180 of human GO Protein alpha (NP_066268.1).,, Sequence:, GFSGEDVKQYKPVVYSNTIQSLAAIVRAMDTLGIEYGDKERKADAKMVCDVVSRMEDTEPFSAELLSAMMRLWGDSGIQECFNRSREYQLNDSAKYYLDSLDRIGAADYQPTEQDILRTRV |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?