missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNL3 Monoclonal Antibody (OTI6F4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
4065.15 NOK - 6057.09 NOK
Specifications
Antigen | GNL3 |
---|---|
Clone | OTI6F4 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17558134
|
Origene Technologies
TA813296S |
30 μL |
4065.15 NOK
30µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17538134
|
Origene Technologies
TA813296 |
100 μL |
6057.09 NOK
100µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
GNL3 Monoclonal antibody specifically detects GNL3 in Human samples. It is validated for Western BlotSpecifications
GNL3 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
C77032; E2IG3; E2-induced gene 3 protein; estradiol-induced nucleotide binding protein; G protein nucleolar 3; Gnl3; guanine nucleotide binding protein-like 3 (nucleolar); guanine nucleotide-binding protein-like 3; MGC800; NNP47; novel nucleolar protein 47; Ns; nucleolar GTP-binding protein 3; Nucleostemin | |
Full length human recombinant protein of human GNL3 (NP_055181) produced in E.coli. | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
GNL3 |
OTI6F4 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9BVP2 | |
26354 | |
IgG1 | |
Protein A/G | |
Antibody |