missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNL3 Monoclonal Antibody (OTI6F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813295
This item is not returnable.
View return policy
Description
GNL3 Monoclonal antibody specifically detects GNL3 in Human samples. It is validated for Western BlotSpecifications
| GNL3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BVP2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 26354 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI6F1 | |
| Unconjugated | |
| GNL3 | |
| C77032; E2IG3; E2-induced gene 3 protein; estradiol-induced nucleotide binding protein; G protein nucleolar 3; Gnl3; guanine nucleotide binding protein-like 3 (nucleolar); guanine nucleotide-binding protein-like 3; MGC800; NNP47; novel nucleolar protein 47; Ns; nucleolar GTP-binding protein 3; Nucleostemin | |
| Full length human recombinant protein of human GNL3 (NP_055181) produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction